N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-D-cysteine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Nα-Fmoc-S-trityl-D-cysteine, Fmoc-D-Cys(Trt)-OH, N-Fmoc-S-trityl-D-cysteine
A230429-10G
A230429-1G
A230429-25G
A230429-5G
A230429-100G
A230429-500G
103850-444EA
10
USD
103850-444
77000-180
77000-182
77303-874
77303-876
77550-492
77672-044
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-D-cysteine 97%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-D-cysteine
Precaución: for Research Use Only
Formula:
C₃₇H₃₁NO₄S MW: 585.72 g/mol Boiling Pt: 763.4 °C (760 mm Hg) Melting Pt: 168…173 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00151922 CAS Number: 167015-11-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...