Fmoc-D-Cha-OH 97%

Supplier: Ambeed
Warning

Synonyms: N-Fmoc-3-cyclohexyl-D-alanine, Fmoc-β-cyclohexyl-D-Ala-OH

A352307-25G A352307-100G A352307-250MG A352307-1G
103826-064EA 10 USD
103826-064 103837-064 76840-174 77155-002 77155-004 77306-630
Fmoc-D-Cha-OH 97%
Fmoc-D-Cha-OH

Precaución: for Research Use Only

Formula: C₂₄H₂₇NO₄
MW: 393.483 g/mol
Boiling Pt: 610.5 °C (760 mm Hg)
Flash Pt: 323 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00151912
CAS Number: 144701-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR