Fmoc-d-asp-otbu 96%

Supplier: Ambeed
Warning

Synonyms: N-Fmoc-D-aspartic acid 1-tert-butyl ester, Fmoc-D-Asp-OtBu

A201604-100G A201604-10G A201604-1G A201604-25G A201604-5G
103817-310EA 11 USD
103817-310 103847-952 76800-378 76800-380 77302-770 77302-772 77302-774
Fmoc-d-asp-otbu 96%
Fmoc-d-asp-otbu

Precaución: for Research Use Only

Formula: C₂₃H₂₅NO₆
Boiling Pt: 617.402 °C (760 mm Hg)
Melting Pt: 90…98 °C
Flash Pt: 327.193 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00237011
CAS Number: 134098-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR