N-Fmoc-D-aspartic acid-4-allyl ester 95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-D-Asp(OAll)-OH

A496422-1G A496422-25G A496422-100G A496422-10G A496422-250MG
103828-258EA 19 USD
103828-258 103839-336 76843-302 76843-304 76991-196 76991-198 76991-200
N-Fmoc-D-aspartic acid-4-allyl ester 95%
N-Fmoc-D-aspartic acid-4-allyl ester

Precaución: for Research Use Only

Formula: C₂₂H₂₁NO₆
MW: 395.411 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01074688
CAS Number: 177609-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR