Fmoc-D-Arg-OH 95%

Supplier: Ambeed
Warning

A121413-100G A121413-10G A121413-250MG A121413-25G A121413-5G
103800-068EA 6 USD
103800-068 77149-188 77149-190 77149-192 77149-194 77312-188
Fmoc-D-Arg-OH 95%
Fmoc-D-Arg-OH

Precaución: for Research Use Only

Formula: C₂₁H₂₄N₄O₄
MW: 396.447 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00077047
CAS Number: 130752-32-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR