Fmoc-D-Arg(Pbf)-OH 97%

Supplier: Ambeed
Warning

Synonyms: Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nomega-(2,2,4,6,7-pentamethyldihydrobenzofuran-5-sulphonyl)-D-arginine, Fmoc-D-Arg(Pbf)-OH, Nα-Fmoc-Nomega-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulphonyl)-D-arginine, Nalpha-Fmoc-Nomega-Pbf-D-arginine

A109500-100G A109500-10G A109500-250MG A109500-25G A109500-1G A109500-5G A109500-500G
103797-514EA 6 USD
103797-514 103823-312 76995-118 76995-120 76995-122 76995-124 77310-128 77310-130 77670-548
Fmoc-D-Arg(Pbf)-OH 97%
Fmoc-D-Arg(Pbf)-OH

Precaución: for Research Use Only

Formula: C₃₄H₄₀N₄O₇S
MW: 648.775 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00237010
CAS Number: 187618-60-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR