Fmoc-Dap-OH 95%

Supplier: Ambeed
Warning

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-aminopropanoic acid

A130208-250MG A130208-5G A130208-1G
103801-394EA 27 USD
103801-394 77094-936 77298-398 77313-826
Fmoc-Dap-OH 95%
Fmoc-Dap-OH

Precaución: for Research Use Only

Formula: C₁₈H₁₈N₂O₄
MW: 326.352 g/mol
Boiling Pt: 579.5 °C (760 mm Hg)
Flash Pt: 304.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00237019
CAS Number: 181954-34-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR