Fmoc-Dap(Boc)-OH 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-(N-beta-boc)-L-alpha,beta-diaminopropionic acid, 3-(tert-Butoxycarbonyl)amino-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-alanine, (S)-3-(tert-Butoxycarbonylamino)-2-[(9H-fluoren-9-ylmethoxy)carbonylamino]propionic acid, Nα-Fmoc-Nβ-Boc-L-2,3-diaminopropionic acid, (S)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid, 3-(Boc-amino)-N-Fmoc-L-alanine

A117408-250MG A117408-1G A117408-5G A117408-10G
103798-882EA 9 USD
103798-882 103824-766 76781-482 77311-546 77311-548 77571-038
Fmoc-Dap(Boc)-OH 97%
Fmoc-Dap(Boc)-OH

Precaución: for Research Use Only

Formula: C₂₃H₂₆N₂O₆
MW: 426.47 g/mol
Boiling Pt: 652.5 °C (760 mm Hg)
Melting Pt: 131…139 °C
Flash Pt: 348.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00235896
CAS Number: 162558-25-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR