Fmoc-D-allo-isoleucine 98%

Supplier: Ambeed
Warning

A224321-1G A224321-5G A224321-10G
103819-098EA 27 USD
103819-098 103849-988 77088-634 77088-636 77152-762 77152-764
Fmoc-D-allo-isoleucine 98%
Fmoc-D-allo-isoleucine

Precaución: for Research Use Only

Formula: C₂₁H₂₃NO₄
Boiling Pt: 559.8 °C (760 mm Hg)
Flash Pt: 292.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00077062
CAS Number: 118904-37-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR