Fmoc-Dab-OH 98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-(Fmoc-amino)-4-aminobutyric acid

A131807-250MG A131807-1G A131807-25G A131807-5G A131807-100G
103801-636EA 16 USD
103801-636 77095-008 77314-162 77314-164 77314-166 77549-568
Fmoc-Dab-OH 98%
Fmoc-Dab-OH

Precaución: for Research Use Only

Formula: C₁₉H₂₀N₂O₄
Boiling Pt: 593.6 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00237017
CAS Number: 161420-87-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR