Fmoc-Dab(Alloc)-OH 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Dab(Aloc)-OH

A197945-1G A197945-25G A197945-5G A197945-10G A197945-250MG
103816-980EA 34 USD
103816-980 103847-606 77085-618 77085-620 77085-622 77097-312 77097-314
Fmoc-Dab(Alloc)-OH 98%
Fmoc-Dab(Alloc)-OH

Precaución: for Research Use Only

Formula: C₂₃H₂₄N₂O₆
MW: 424.453 g/mol
Boiling Pt: 676.7 °C (760 mm Hg)
Flash Pt: 363.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00798634
CAS Number: 204316-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR