N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-L-cysteine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Fmoc-Cys(Trt)-OH, N-Fmoc-S-trityl-L-cysteine
A245081-25G
A245081-5G
A245081-1000G
A245081-100G
A245081-1G
A245081-500G
A245081-10G
103851-456EA
6
USD
103851-456
77090-690
77090-692
77276-134
77276-136
77276-138
77276-140
77593-558
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-L-cysteine 98%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-L-cysteine
Precaución: for Research Use Only
Formula:
C₃₇H₃₁NO₄S Boiling Pt: 763.4 °C (760 mm Hg) Melting Pt: 172…177 °C Flash Pt: 415.5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00038538 CAS Number: 103213-32-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...