Fmoc-Cys(tBu)-OH 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-Fmoc-S-(tert-butyl)-L-cysteine, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(tert-butyl)-L-cysteine, N-Fmoc-S-tert-butyl-L-cysteine, Fmoc-Cys(tBu)-OH
A163032-100G
A163032-10G
A163032-250MG
A163032-1G
A163032-25G
A163032-5G
103805-678EA
6
USD
103805-678
103843-936
77286-522
77286-524
77286-526
77300-624
77300-626
77300-628
Fmoc-Cys(tBu)-OH 98%
Fmoc-Cys(tBu)-OH
Precaución: for Research Use Only
Formula:
C₂₂H₂₅NO₄S MW: 399.505 g/mol Boiling Pt: 603.4 °C (760 mm Hg) Melting Pt: 135…137 °C Flash Pt: 318.7 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00037130 CAS Number: 67436-13-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...