Fmoc-Cys(pMeOBzl)-OH 98%

Supplier: Ambeed
Warning

Synonyms: N-Fmoc-S-(4-methoxybenzyl)-L-cysteine, Fmoc-Cys(Mob)-OH

A170933-1G A170933-5G A170933-100G
103806-560EA 14 USD
103806-560 103844-836 77081-956 77081-958 77881-317
Fmoc-Cys(pMeOBzl)-OH 98%
Fmoc-Cys(pMeOBzl)-OH

Precaución: for Research Use Only

Formula: C₂₆H₂₅NO₅S
MW: 463.548 g/mol
Boiling Pt: 689.5 °C (760 mm Hg)
Melting Pt: 140 °C
Flash Pt: 370.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00065635
CAS Number: 141892-41-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR