Fmoc-Cys(Et)-OH 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-Fmoc-S-ethyl-L-cysteine
A101966-250MG
A101966-1G
103796-230EA
22
USD
103796-230
103821-928
76796-294
77308-810
Fmoc-Cys(Et)-OH 95%
Fmoc-Cys(Et)-OH
Precaución: for Research Use Only
Formula:
C₂₀H₂₁NO₄S Storage Temperature: Refrigerator |
MDL Number:
MFCD00151921 CAS Number: 200354-34-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...