Fmoc-Cys(Dpm)-OH 97%

Supplier: Ambeed
Warning

A521423-1G A521423-10G A521423-250MG A521423-100G
103828-510EA 6 USD
103828-510 103839-618 76843-620 77004-846 77004-848 77882-275
Fmoc-Cys(Dpm)-OH 97%
Fmoc-Cys(Dpm)-OH

Precaución: for Research Use Only

Formula: C₃₁H₂₇NO₄S
MW: 509.63 g/mol
Boiling Pt: 714.3±60.0 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD02682541
CAS Number: 247595-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR