(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-cyclopentylacetic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Fmoc-Cpg-OH, N-Fmoc-2-cyclopentyl-L-glycine
A154887-100MG
A154887-1G
A154887-10G
A154887-100G
A154887-250MG
103804-688EA
23
USD
103804-688
103842-882
76798-774
77300-084
77567-260
77667-192
77879-072
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-cyclopentylacetic acid 98%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-cyclopentylacetic acid
Precaución: For research use only.
Formula:
C₂₂H₂₃NO₄ MW: 365.429 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD01311751 CAS Number: 220497-61-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...