Fmoc-(S)-3-Cyclopentylalanine 97%

Supplier: Ambeed
Warning

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-cyclopentylpropanoic acid

A248147-100MG A248147-250MG A248147-5G
103811-812EA 57 USD
103811-812 103851-648 77013-926 77013-928 77090-984
Fmoc-(S)-3-Cyclopentylalanine 97%
Fmoc-(S)-3-Cyclopentylalanine

Precaución: for Research Use Only

Formula: C₂₃H₂₅NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD03094886
CAS Number: 371770-32-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR