Fmoc-Chg-OH 97%
Supplier: Ambeed
|
Warning
|
A317729-25G
A317729-5G
A317729-10G
103836-078EA
7
USD
103836-078
76838-924
76838-926
77099-780
Fmoc-Chg-OH 97%
Fmoc-Chg-OH
Precaución: for Research Use Only
Formula:
C₂₃H₂₅NO₄ Boiling Pt: 602.8 °C (760 mm Hg) Flash Pt: 318.409 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00190878 CAS Number: 161321-36-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...