N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-cyclohexyl-L-alanine 97%

Supplier: Ambeed
Warning

A751063-25G A751063-5G A751063-100G A751063-10G A751063-250MG A751063-1G
103840-850EA 17 USD
103840-850 76845-086 76845-088 77158-886 77158-888 77158-890 77308-250
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-cyclohexyl-L-alanine 97%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-cyclohexyl-L-alanine

Precaución: for Research Use Only

Formula: C₂₄H₂₇NO₄
Boiling Pt: 610.5 °C (760 mm Hg)
Flash Pt: 323 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00065614
CAS Number: 135673-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR