Fmoc-Bpa-OH 95%

Supplier: Ambeed
Warning

Synonyms: N-Fmoc-4-benzoyl-L-phenylalanine, Fmoc-p-Bz-Phe-OH

A108761-1G A108761-5G
103797-386EA 46 USD
103797-386 103823-172 77283-774 77310-002
Fmoc-Bpa-OH 95%
Fmoc-Bpa-OH

Precaución: for Research Use Only

Formula: C₃₁H₂₅NO₅
MW: 491.543 g/mol
Boiling Pt: 729 °C (760 mm Hg)
Flash Pt: 394.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00151937
CAS Number: 117666-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR