Fmoc-L-4,4'-biphenylalanine 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-4-phenyl-Phe-OH, Fmoc-Bip(4,4')-OH

A112946-100G A112946-100MG A112946-10G A112946-250MG A112946-25G A112946-1G
103798-136EA 15 USD
103798-136 103823-970 76995-316 76995-318 76995-320 76995-322 76995-324 77310-806
Fmoc-L-4,4'-biphenylalanine 98%
Fmoc-L-4,4'-biphenylalanine

Precaución: for Research Use Only

Formula: C₃₀H₂₅NO₄
Boiling Pt: 700.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00191198
CAS Number: 199110-64-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR