(S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: N-Fmoc-L-aspartic Acid 1-tert-Butyl Ester, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic Acid 1-tert-Butyl Ester, Fmoc-Asp(OH)-OtBu, Fmoc-Asp-OtBu, 1-tert-Butyl N-Fmoc-L-aspartate

A193717-100G A193717-10G A193717-250MG A193717-500G A193717-1G A193717-25G A193717-5G
103816-574EA 9 USD
103816-574 103847-188 76785-026 76785-028 76785-030 76785-032 77085-090 77302-358 77302-360
(S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid 98%
(S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid

Precaución: for Research Use Only

Formula: C₂₃H₂₅NO₆
Boiling Pt: 617.4 °C (760 mm Hg)
Flash Pt: 327.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00065631
CAS Number: 129460-09-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR