Fmoc-Asp-OBzl 98%

Supplier: Ambeed
Warning

Synonyms: N-Fmoc-L-aspartic acid 1-benzyl ester, Fmoc-Asp-Obzl

A176288-1G A176288-5G A176288-10G A176288-25G A176288-100G
103807-130EA 10 USD
103807-130 103845-450 77082-800 77082-802 77615-474 77615-476 77748-151
Fmoc-Asp-OBzl 98%
Fmoc-Asp-OBzl

Precaución: for Research Use Only

Formula: C₂₆H₂₃NO₆
MW: 445.471 g/mol
Boiling Pt: 683.7 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD00198201
CAS Number: 86060-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR