(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: N-Fmoc-L-aspartic Acid 4-tert-Butyl Ester, 4-tert-Butyl N-Fmoc-L-aspartate, Fmoc-Asp(OtBu)-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic Acid 4-tert-Butyl Ester

A131522-100G A131522-10G A131522-1G A131522-500G A131522-25G A131522-1000G
103808-428EA 415 USD
103808-428 77272-018 77272-020 77272-022 77272-024 77314-096 77747-938
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid 98%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid

Precaución: for Research Use Only

Formula: C₂₃H₂₅NO₆
Boiling Pt: 620.8 °C (760 mm Hg)
Melting Pt: 148…150 °C
Flash Pt: 329.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00037131
CAS Number: 71989-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR