Fmoc-aspartic acid-4-methyl ester 98%

Supplier: Ambeed
Warning

Synonyms: N-Fmoc-L-aspartic acid 4-methyl ester, Fmoc-Asp(OMe)-OH

A203046-1G A203046-25G A203046-5G A203046-10G A203046-100G A203046-500G
103817-434EA 5 USD
103817-434 103848-088 77086-204 77086-206 77086-208 77567-942 77611-970 77883-470
Fmoc-aspartic acid-4-methyl ester 98%
Fmoc-aspartic acid-4-methyl ester

Precaución: For research use only.

Formula: C₂₀H₁₉NO₆
Boiling Pt: 613.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08458573
CAS Number: 145038-53-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR