Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-asparagine 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Asn-OH, Nɑ-Fmoc-L-asparagine

A120859-100G A120859-10G A120859-25G A120859-5G
76766-456EA 44 USD
76766-456 76766-458 76766-460 77297-798
Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-asparagine 98%
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-asparagine

Precaución: for Research Use Only

Formula: C₁₉H₁₈N₂O₅
Boiling Pt: 678.6 °C (760 mm Hg)
Melting Pt: 190 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037132
CAS Number: 71989-16-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR