Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-γ-trityl-L-asparagine 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Asn(Trt)-OH, Nɑ-Fmoc-Nγ-trityl-L-asparagine

A424990-1G A424990-25G A424990-5G A424990-100G A424990-10G A424990-500G A424990-1000G
103827-454EA 7 USD
103827-454 103838-504 76842-152 76842-154 76842-156 77279-012 77279-014 77279-016 77616-842
Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-γ-trityl-L-asparagine 98%
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-gamma-trityl-L-asparagine

Precaución: for Research Use Only

Formula: C₃₈H₃₂N₂O₅
Melting Pt: 201…204 °C
Flash Pt: 472.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00077049
CAS Number: 132388-59-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR