Fmoc-Arg(Pbf)-OH 98%

Supplier: Ambeed
Warning

A116775-100G A116775-10G A116775-25G A116775-5G A116775-1000G A116775-500G
103824-662EA 7 USD
103824-662 77271-246 77271-248 77271-250 77311-460 77549-336 77549-338
Fmoc-Arg(Pbf)-OH 98%
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nomega-(2,2,4,6,7-pentamethyldihydrobenzofuran-5-sulfonyl)-L-arginine

Precaución: for Research Use Only

Formula: C₃₄H₄₀N₄O₇S
Melting Pt: 132 °C
Storage Temperature: Freezer
MDL Number: MFCD00235804
CAS Number: 154445-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR