Fmoc-allo-Thr-OH 95%
Supplier: Ambeed
|
Warning
|
A168059-250MG
A168059-1G
A168059-5G
A168059-25G
103806-242EA
23
USD
103806-242
103844-500
76799-288
77081-538
77081-540
77879-127
Fmoc-allo-Thr-OH 95%
Fmoc-allo-Thr-OH
Precaución: for Research Use Only
Formula:
C₁₉H₁₉NO₅ Melting Pt: 152…156 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01318739 CAS Number: 146306-75-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...