Fmoc-allo-Thr(tBu)-OH 97%

Supplier: Ambeed
Warning

Synonyms: N-Fmoc-O-tert-butyl-L-allo-threonine, Fmoc-Allo-Thr(tBu)-OH

A118672-100MG A118672-250MG A118672-1G A118672-5G A118672-10G A118672-25G
103799-094EA 18 USD
103799-094 76797-238 76797-240 77297-620 77297-622 77566-176 77566-178
Fmoc-allo-Thr(tBu)-OH 97%
Fmoc-allo-Thr(tBu)-OH

Precaución: For research use only.

Formula: C₂₃H₂₇NO₅
MW: 397.471 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00237371
CAS Number: 201481-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR