8-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-3,6-dioxa-n-octanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-NH-PEG2-CH2COOH, 8-(Fmoc-amino)-3,6-dioxa-n-octanoic acid, Fmoc-8-amino-3,6-dioxaoctanoic acid, [2-[2-[(Fmoc-amino)ethoxy]ethoxy]acetic acid, 1-(9H-Fluoren-9-yl)-3-oxo-2,7,10-trioxa-4-azadodecan-12-oic acid, [2-[2-[(9H-Fluoren-9-ylmethoxy)carbonylamino]ethoxy]ethoxy]acetic acid

A315456-100G A315456-250MG A315456-1G A315456-10G A315456-500G
103825-072EA 10 USD
103825-072 103835-996 76802-796 76802-798 76838-824 77550-876 77550-878
8-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-3,6-dioxa-n-octanoic acid 98%
8-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-3,6-dioxa-n-octanoic acid

Precaución: for Research Use Only

Formula: C₂₁H₂₃NO₆
MW: 385.42 g/mol
Boiling Pt: 631.4 °C (760 mm Hg)
Flash Pt: 335.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01321015
CAS Number: 166108-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR