Fmoc-Abu-OH 98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-(Fmoc-amino)butyric acid

A103086-100G A103086-10G A103086-25G A103086-5G A103086-500G
103822-182EA 7 USD
103822-182 76994-628 76994-630 76994-632 77309-054 77670-486
Fmoc-Abu-OH 98%
Fmoc-Abu-OH

Precaución: for Research Use Only

Formula: C₁₉H₁₉NO₄
MW: 325.364 g/mol
Boiling Pt: 550.7±33.0 °C (760 mm Hg)
Melting Pt: 130…134 °C
Flash Pt: 286.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00080268
CAS Number: 135112-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR