Fmoc-5-Hydroxy-L-tryptophan 95%

Supplier: Ambeed
Warning

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(5-hydroxy-1H-indol-3-yl)propanoic acid, N-Fmoc-5-hydroxy-L-tryptophan, Fmoc-Trp(5-OH)-OH

A114612-100MG A114612-250MG A114612-5G
103798-430EA 17 USD
103798-430 103824-270 77271-126 77271-128 77311-088
Fmoc-5-Hydroxy-L-tryptophan 95%
Fmoc-5-Hydroxy-L-tryptophan

Precaución: for Research Use Only

Formula: C₂₆H₂₂N₂O₅
MW: 442.46 g/mol
Boiling Pt: 760.3±60.0 °C (760 mm Hg)
Flash Pt: 413.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00270744
CAS Number: 178119-94-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR