Fmoc-5-Chloro-L-tryptophan 95%

Supplier: Ambeed
Warning

A183935-100MG A183935-250MG A183935-1G A183935-5G
103815-690EA 125 USD
103815-690 103846-252 77029-556 77029-558 77083-864 77083-866
Fmoc-5-Chloro-L-tryptophan 95%
Fmoc-5-Chloro-L-tryptophan

Precaución: for Research Use Only

Formula: C₂₆H₂₁ClN₂O₄
Boiling Pt: 732.6±60.0 °C (760 mm Hg)
Flash Pt: 396.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16883066
CAS Number: 1257849-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR