Fmoc-3-(3-pyridyl)-L-alanine 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-3-Pal-OH, Fmoc-β-(3-pyridyl)-Ala-OH

A168107-10G A168107-250MG A168107-1G A168107-5G
103806-252EA 10 USD
103806-252 103844-510 76784-126 76784-128 77081-556 77081-558
Fmoc-3-(3-pyridyl)-L-alanine 97%
Fmoc-3-(3-pyridyl)-L-alanine

Precaución: for Research Use Only

Formula: C₂₃H₂₀N₂O₄
MW: 388.423 g/mol
Boiling Pt: 645.6 °C (760 mm Hg)
Melting Pt: 155.3 °C
Flash Pt: 344.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00144887
CAS Number: 175453-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR