Fmoc-Thi-OH, Fmoc-3-L-Ala(3-Thienyl)-OH 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Thien-3-yl-L-alanine, N-FMOC protected
A657694-100MG
A657694-250MG
A657694-1G
A657694-5G
103840-424EA
510
USD
103840-424
76727-394
76727-396
76727-398
77308-070
77308-072
Fmoc-Thi-OH, Fmoc-3-L-Ala(3-Thienyl)-OH 97%
Fmoc-Thi-OH, Fmoc-3-L-Ala(3-Thienyl)-OH
Precaución: for Research Use Only
Formula:
C₂₂H₁₉NO₄S MW: 393.4 g/mol Boiling Pt: 626.3 °C (760 mm Hg) Melting Pt: 188.4 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00151916 CAS Number: 186320-06-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...