N-Fmoc-3-(2-pyridyl)-L-alanine 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-2-Pal-OH

A139664-10G A139664-250MG A139664-1G A139664-25G A139664-5G
103802-762EA 68 USD
103802-762 76996-820 76996-822 77299-082 77299-084 77315-858
N-Fmoc-3-(2-pyridyl)-L-alanine 97%
N-Fmoc-3-(2-pyridyl)-L-alanine

Precaución: for Research Use Only

Formula: C₂₃H₂₀N₂O₄
MW: 388.423 g/mol
Boiling Pt: 630.9±55.0 °C (760 mm Hg)
Melting Pt: 151 °C
Flash Pt: 335.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00672564
CAS Number: 185379-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR