Fmoc-3-(2-naphthyl)-L-alanine 95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-2-Nal-OH

A263589-10G A263589-250MG A263589-500G A263589-1G A263589-25G A263589-5G A263589-100G
103812-630EA 7 USD
103812-630 103852-518 77000-994 77000-996 77000-998 77092-154 77092-156 77092-158 77571-628
Fmoc-3-(2-naphthyl)-L-alanine 95%
Fmoc-3-(2-naphthyl)-L-alanine

Precaución: for Research Use Only

Formula: C₂₈H₂₃NO₄
MW: 437.495 g/mol
Flash Pt: 371.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00144886
CAS Number: 112883-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR