Fmoc-β-(2-pyridyl)-D-alanine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Fmoc-2-D-Pal-OH
A376796-1G
A376796-10G
A376796-250MG
103826-594EA
42
USD
103826-594
103837-620
76840-924
77003-588
77003-590
Fmoc-β-(2-pyridyl)-D-alanine 97%
Fmoc-beta-(2-pyridyl)-D-alanine
Precaución: for Research Use Only
Formula:
C₂₃H₂₀N₂O₄ Boiling Pt: 630.9 °C (760 mm Hg) Melting Pt: 154.5 °C Flash Pt: 335.4 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00672565 CAS Number: 185379-39-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...