(Fmoc-amino)-PEG6-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-N-amido-dPEG6-acid, 1-(9H-Fluoren-9-yl)-3-oxo-2,7,10,13,16,19,22-heptaoxa-4-azapentacosan-25-oic acid, Carboxy-PEG6-(Fmoc-amine), Fmoc-21-amino-4,7,10,13,16,19-hexaoxaheneicosanoic acid, 21-(Fmoc-amino)-4,7,10,13,16,19-hexaoxahenicosanoic acid, 21-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-4,7,10,13,16,19-hexaoxahenicosanoic acid, Fmoc-N-amido-PEG6-acid

A356631-100MG A356631-250MG A356631-25G A356631-5G A356631-1G
103826-174EA 360 USD
103826-174 76648-526 76648-528 76648-530 76648-532 76840-322
(Fmoc-amino)-PEG6-carboxylic acid 95%
(Fmoc-amino)-PEG6-carboxylic acid

Precaución: for Research Use Only

Formula: C₃₀H₄₁NO₁₀
Boiling Pt: 734.8±60.0 °C (760 mm Hg)
Flash Pt: 398.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06656474
CAS Number: 882847-34-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR