2-Fluoro-α-methyl-4-biphenylacetic acid 98%

Supplier: Ambeed
Danger

Synonyms: Ansaid, 2-(2-Fluoro-[1,1'-biphenyl]-4-yl)propanoic acid, Flurbiprofen, Froben, Antadys

A481951-100G A481951-100MG A481951-25G A481951-5G A481951-1G A481951-10G
103828-136EA 18 USD
103828-136 103839-210 76804-832 76804-834 76843-154 76843-156 77307-538 77602-086
2-Fluoro-α-methyl-4-biphenylacetic acid 98%
2-Fluoro-α-methyl-4-biphenylacetic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₃FO₂
MW: 244.265 g/mol
Melting Pt: 110…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00079303
CAS Number: 5104-49-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR