Fluoromethyl 4-methylbenzenesulfonate 97%

Supplier: Ambeed
Danger

A276228-100MG A276228-250MG
103813-300EA 58 USD
103813-300 76827-420 76827-422
Fluoromethyl 4-methylbenzenesulfonate 97%
Fluoromethyl 4-methylbenzenesulfonate

Precaución: for Research Use Only

Formula: C₈H₉FO₃S
MW: 204.219 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20482732
CAS Number: 114435-86-8
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR