Flunixin meglumine 98%

Supplier: Ambeed
Warning

Synonyms: (2R,3R,4R,5S)-6-(Methylamino)hexane-1,2,3,4,5-pentaol 2-((2-methyl-3-(trifluoromethyl)phenyl)amino)nicotinate

A101184-100G A101184-250MG A101184-25G A101184-5G
103795-842EA 7 USD
103795-842 76796-258 76796-260 76796-262 76796-264
Flunixin meglumine 98%
Flunixin meglumine

Precaución: for Research Use Only

Formula: C₁₄H₁₁F₃N₂O₂·C₇H₁₇NO₅
MW: 491.4581 g/mol
Boiling Pt: 378.7 °C (760 mm Hg)
Melting Pt: 136…138 °C
Flash Pt: 182.8 °C
Storage Temperature: Ambient
MDL Number: MFCD01725419
CAS Number: 42461-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR