Fludarabine monophosphate 98%

Supplier: Ambeed
Warning

Synonyms: ((2R,3S,4S,5R)-5-(6-Amino-2-fluoro-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl dihydrogen phosphate, Fludarabine Phosphate, 2-F-ara-AMP, 9-(beta-D-Arabinofuranosyl)-2-fluoroadenine 5'-Monophosphate, 2-Fluoroadenine 9-beta-D-Arabinofuranoside 5'-Monophosphate

A103732-10MG A103732-1MG A103732-250MG A103732-50MG A103732-5G
103796-604EA 84 USD
103796-604 103822-314 77107-372 77107-374 77107-376 77107-378 77283-156
Fludarabine monophosphate 98%
Fludarabine monophosphate

Precaución: for Research Use Only

Formula: C₁₀H₁₃FN₅O₇P
MW: 365.21 g/mol
Melting Pt: >155 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00866418
CAS Number: 75607-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR