Flubendazole 98%

Supplier: Ambeed
Warning

A100488-100MG A100488-25MG
103795-472EA 24 USD
103795-472 103821-166 76796-226 76796-228
Flubendazole 98%
Flubendazole

Precaución: for Research Use Only

Formula: C₁₆H₁₂FN₃O₃
Melting Pt: 290 °C
Storage Temperature: Ambient
MDL Number: MFCD00871999
CAS Number: 31430-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR