5-Fluoro-2'-deoxyuridine (floxuridine) 98%

Supplier: Ambeed
Danger

A230277-1G A230277-250MG A230277-1MG
103819-532EA 14 USD
103819-532 103850-430 77089-290 77249-554 77249-556 77746-345
5-Fluoro-2'-deoxyuridine (floxuridine) 98%
5-Fluoro-2'-deoxyuridine (floxuridine)

Precaución: for Research Use Only

Formula: C₉H₁₁FN₂O₅
Melting Pt: 148 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006530
CAS Number: 50-91-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR