Firocoxib 99%

Supplier: Ambeed
Warning

Synonyms: 3-(Cyclopropylmethoxy)-5,5-dimethyl-4-(4-(methylsulphonyl)phenyl)furan-2(5H)-one, ML1785713, 3-(Cyclopropylmethoxy)-4-(4-methylsulphonylphenyl)-5,5-dimethylfuranone, Firocoxib

A196334-100MG A196334-1MG A196334-2MG A196334-5MG A196334-10MG A196334-50MG A196334-1G A196334-5G A196334-250MG
103792-358EA 85 USD
103792-358 103799-464 76873-236 76873-238 76873-240 76873-242 77085-410 77085-412 77611-888 77611-890 77671-744
Firocoxib 99%
Firocoxib

Precaución: for Research Use Only

Formula: C₁₇H₂₀O₅S
MW: 336.40 g/mol
Boiling Pt: 563.9 °C (760 mm Hg)
Melting Pt: 78…80 °C
Flash Pt: 294.8 °C
Storage Temperature: Freezer
MDL Number: MFCD09837800
CAS Number: 189954-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR