Fexinidazole 98%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-2-((4-(methylthio)phenoxy)methyl)-5-nitro-1H-imidazole, HOE 239

A322155-100MG A322155-1MG A322155-250MG A322155-50MG A322155-10MG A322155-1G A322155-5MG
103791-626EA 14 USD
103791-626 103792-612 103825-296 76802-912 76802-914 76802-916 76802-918 76839-144 76839-146 76839-148
Fexinidazole 98%
Fexinidazole

Precaución: for Research Use Only

Formula: C₁₂H₁₃N₃O₃S
MW: 279.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00866607
CAS Number: 59729-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR