Fasudil hydrochloride 98%
Supplier: Ambeed
|
Warning
|
A193859-100MG
A193859-1MG
A193859-250MG
A193859-1G
A193859-25G
A193859-100G
103816-592EA
64
USD
103816-592
103847-200
76800-076
76800-078
76800-080
77085-102
77667-620
77879-232
Fasudil hydrochloride 98%
Fasudil hydrochloride
Precaución: for Research Use Only
Formula:
C₁₄H₁₇N₃O₂S Flash Pt: 259.9 °C Storage Temperature: Freezer |
MDL Number:
MFCD00943198 CAS Number: 105628-07-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...